COM490021461 Combi-Blocks, Inc.

2,4-Decadien-1-ol

  • MDL number MFCD00014052
  • Cas Number : 14507-02-9
  • Molecular Weight : 154.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7988

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7988
Availability details for
Quantity
  • Availability
  • Add to Cart