COM490021823 Combi-Blocks, Inc.

2-bromo-4-fluoro-1-methoxy-3-nitrobenzene

  • MDL number MFCD28739833
  • Cas Number : 1805420-95-4
  • Molecular Weight : 250.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YF-0681

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YF-0681
Availability details for
Quantity
  • Availability
  • Add to Cart