COM490021828 Combi-Blocks, Inc.

1-Bromo-3-fluoro-4-methoxy-2-nitrobenzene

  • MDL number MFCD28739842
  • Cas Number : 1807044-38-7
  • Molecular Weight : 250.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YF-0687

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YF-0687
Availability details for
Quantity
  • Availability
  • Add to Cart