COM490021995 Combi-Blocks, Inc.

Methyl 2-methoxy-4-nitrobenzoate

  • MDL number MFCD11041296
  • Cas Number : 39106-79-1
  • Molecular Weight : 211.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YF-0966

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YF-0966
Availability details for
Quantity
  • Availability
  • Add to Cart