COM490021996 Combi-Blocks, Inc.

Methyl 3-(3-nitrophenyl)propanoate

  • MDL number MFCD22987434
  • Cas Number : 22768-05-4
  • Molecular Weight : 209.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YF-0967

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YF-0967
Availability details for
Quantity
  • Availability
  • Add to Cart