COM490022648 Combi-Blocks, Inc.

5-Bromo-1-isobutyl-1H-pyrazole

  • MDL number MFCD23134839
  • Cas Number : 1427012-49-4
  • Molecular Weight : 203.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-1-isobutyl-1H-pyrazole
Inchi Code 1S/C7H11BrN2/c1-6(2)5-10-7(8)3-4-9-10/h3-4,6H,5H2,1-2H3
Inchi Key YUWRPYZOXLGDTB-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YF-2152

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YF-2152
Availability details for
Quantity
  • Availability
  • Add to Cart