COM490022650 Combi-Blocks, Inc.

1-bromo-4,4-difluorocyclohex-1-ene

  • MDL number MFCD29043775
  • Cas Number : 1881332-54-2
  • Molecular Weight : 197.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YF-2183

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YF-2183
Availability details for
Quantity
  • Availability
  • Add to Cart