COM490022654 Combi-Blocks, Inc.

5-Bromo-2-(ethylsulfanyl)pyridine

  • MDL number MFCD14650168
  • Cas Number : 127800-66-2
  • Molecular Weight : 218.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YF-2216

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YF-2216
Availability details for
Quantity
  • Availability
  • Add to Cart