COM490022662 Combi-Blocks, Inc.

5-Bromo-3-methylthiophene-2-carbonitrile

  • MDL number MFCD17169887
  • Cas Number : 38239-48-4
  • Molecular Weight : 202.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YF-2286

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YF-2286
Availability details for
Quantity
  • Availability
  • Add to Cart