COM490022753 Combi-Blocks, Inc.

1,5-dibromo-2-fluoro-3-methoxybenzene

  • MDL number MFCD28784582
  • Cas Number : 1803836-83-0
  • Molecular Weight : 283.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YF-2686

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YF-2686
Availability details for
Quantity
  • Availability
  • Add to Cart