COM490022779 Combi-Blocks, Inc.

5-Chloro-1,3-dibromo-2-fluorobenzene

  • MDL number MFCD28166159
  • Cas Number : 1805525-99-8
  • Molecular Weight : 288.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YF-2786

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YF-2786
Availability details for
Quantity
  • Availability
  • Add to Cart