COM490022907 Combi-Blocks, Inc.

3-Bromo-2-methoxybenzotrifluoride

  • MDL number MFCD13194485
  • Cas Number : 871352-91-9
  • Molecular Weight : 255.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YF-3094

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YF-3094
Availability details for
Quantity
  • Availability
  • Add to Cart