COM490022916 Combi-Blocks, Inc.

1-Bromo-2,3,4-trichlorobenzene

  • MDL number MFCD01026473
  • Cas Number : 81067-37-0
  • Molecular Weight : 260.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YF-3130

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YF-3130
Availability details for
Quantity
  • Availability
  • Add to Cart