COM490023165 Combi-Blocks, Inc.

4-Bromo-n-(4-chlorophenyl)benzamide

  • MDL number MFCD00588671
  • Cas Number : 21354-23-4
  • Molecular Weight : 310.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YF-3766

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YF-3766
Availability details for
Quantity
  • Availability
  • Add to Cart