COM490023167 Combi-Blocks, Inc.

3-Bromo-n-(2-bromophenyl)benzamide

  • MDL number MFCD00587814
  • Cas Number : 346718-81-8
  • Molecular Weight : 355.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YF-3770

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YF-3770
Availability details for
Quantity
  • Availability
  • Add to Cart