COM490023572 Combi-Blocks, Inc.

1-Chloro-2,4-difluoro-5-methoxybenzene

  • MDL number MFCD18415547
  • Cas Number : 1261628-93-6
  • Molecular Weight : 178.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-chloro-2,4-difluoro-5-methoxybenzene
Inchi Code 1S/C7H5ClF2O/c1-11-7-2-4(8)5(9)3-6(7)10/h2-3H,1H3
Inchi Key FZDCQMDEJJMLAZ-UHFFFAOYSA-N

Description

Purity 96%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YF-4858

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YF-4858
Availability details for
Quantity
  • Availability
  • Add to Cart