COM490023734 Combi-Blocks, Inc.

1-Bromo-4-chloronaphthalene

  • MDL number MFCD00971829
  • Cas Number : 53220-82-9
  • Molecular Weight : 241.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YF-5148

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YF-5148
Availability details for
Quantity
  • Availability
  • Add to Cart