COM490023880 Combi-Blocks, Inc.

3-N-Butoxy-4-chlorobenzotrifluoride

  • MDL number MFCD18911712
  • Cas Number : 1443345-23-0
  • Molecular Weight : 252.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YF-5362

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YF-5362
Availability details for
Quantity
  • Availability
  • Add to Cart