COM490023882 Combi-Blocks, Inc.

4-chloro-1-ethoxy-2-(trifluoromethyl)benzene

  • MDL number MFCD29044197
  • Cas Number : 1881293-97-5
  • Molecular Weight : 224.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YF-5364

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YF-5364
Availability details for
Quantity
  • Availability
  • Add to Cart