COM490023984 Combi-Blocks, Inc.

3-Chloro-5-(trifluoromethoxy)benzaldehyde

  • MDL number MFCD06660305
  • Cas Number : 433926-48-8
  • Molecular Weight : 224.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YF-5493

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YF-5493
Availability details for
Quantity
  • Availability
  • Add to Cart