COM490024056 Combi-Blocks, Inc.

Selena sel10607110

  • MDL number MFCD16240854
  • Cas Number : 755024-78-3
  • Molecular Weight : 255.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YF-5583

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YF-5583
Availability details for
Quantity
  • Availability
  • Add to Cart