COM490024399 Combi-Blocks, Inc.

3-(Methoxymethyl)phenol

  • MDL number MFCD14680990
  • Cas Number : 57234-51-2
  • Molecular Weight : 138.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YF-6228

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YF-6228
Availability details for
Quantity
  • Availability
  • Add to Cart