COM490024495 Combi-Blocks, Inc.

4-(2-Bromoethoxy)phenol

  • MDL number MFCD07780181
  • Cas Number : 31406-95-8
  • Molecular Weight : 217.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YF-6476

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YF-6476
Availability details for
Quantity
  • Availability
  • Add to Cart