COM490024771 Combi-Blocks, Inc.

4-[(cyclopentylsulfanyl)methyl]phenol

  • MDL number MFCD29044507
  • Cas Number : 1881293-91-9
  • Molecular Weight : 208.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YF-7001

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YF-7001
Availability details for
Quantity
  • Availability
  • Add to Cart