COM490024931 Combi-Blocks, Inc.

5-(Trifluoromethyl)benzene-1,3-diol

  • MDL number MFCD00061289
  • Cas Number : 454-68-2
  • Molecular Weight : 178.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-(trifluoromethyl)-1,3-benzenediol
Inchi Code 1S/C7H5F3O2/c8-7(9,10)4-1-5(11)3-6(12)2-4/h1-3,11-12H
Inchi Key LXCRBEFECXIJJV-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YF-7253

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YF-7253
Availability details for
Quantity
  • Availability
  • Add to Cart