COM490024943 Combi-Blocks, Inc.

4-Methoxy-2-(trifluoromethyl)phenol

  • MDL number MFCD13185697
  • Cas Number : 53903-50-7
  • Molecular Weight : 192.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YF-7272

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YF-7272
Availability details for
Quantity
  • Availability
  • Add to Cart