COM490025264 Combi-Blocks, Inc.

3-[[(1,1-Dimethylethyl)dimethylsilyl]oxy]-phenol

  • MDL number MFCD27945625
  • Cas Number : 161006-18-4
  • Molecular Weight : 224.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-((tert-butyldimethylsilyl)oxy)phenol
Inchi Code 1S/C12H20O2Si/c1-12(2,3)15(4,5)14-11-8-6-7-10(13)9-11/h6-9,13H,1-5H3
Inchi Key WNTUMICETVLAST-UHFFFAOYSA-N

Description

Purity 96%
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YF-7694

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YF-7694
Availability details for
Quantity
  • Availability
  • Add to Cart