COM497509686 Combi-Blocks, Inc.

3-Chloro-4-fluorophenyl isothiocyanate

  • MDL number MFCD00060700
  • Cas Number : 137724-66-4
  • Molecular Weight : 187.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-4551

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-4551
Availability details for
Quantity
  • Availability
  • Add to Cart