COM497509751 Combi-Blocks, Inc.

2-Amino-6-benzothiazolol, HCl

  • MDL number MFCD04038462
  • Cas Number : 26278-78-4
  • Molecular Weight : 202.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-6055

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-6055
Availability details for
Quantity
  • Availability
  • Add to Cart