COM497509843 Combi-Blocks, Inc.

(4R,5R)-[5-(4-Iodophenyl)-2-phenyl-4,5-dihydro-oxazol-4-yl]methanol

  • MDL number MFCD10687188
  • Cas Number : 927689-68-7
  • Molecular Weight : 379.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8734

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8734
Availability details for
Quantity
  • Availability
  • Add to Cart