COM497510143 Combi-Blocks, Inc.

Ethyl 2-isobutylacetoacetate

  • MDL number MFCD00457371
  • Cas Number : 1522-34-5
  • Molecular Weight : 186.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-9887

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-9887
Availability details for
Quantity
  • Availability
  • Add to Cart