COM497510214 Combi-Blocks, Inc.

Sodium chloroacetate

  • MDL number MFCD00002684
  • Cas Number : 3926-62-3
  • Molecular Weight : 116.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-7029

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-7029
Availability details for
Quantity
  • Availability
  • Add to Cart