COM497510590 Combi-Blocks, Inc.

M-Tolylhydrazine

  • MDL number MFCD00047815
  • Cas Number : 536-89-0
  • Molecular Weight : 122.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-6579

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-6579
Availability details for
Quantity
  • Availability
  • Add to Cart