COM497510637 Combi-Blocks, Inc.

5-Ethoxy-2-mercaptobenzimidazole

  • MDL number MFCD01326487
  • Cas Number : 55489-15-1
  • Molecular Weight : 194.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-7377

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-7377
Availability details for
Quantity
  • Availability
  • Add to Cart