COM497510660 Combi-Blocks, Inc.

1-(2-Chloro-4-nitrophenyl)azepane

  • MDL number MFCD11039792
  • Cas Number : 1260897-00-4
  • Molecular Weight : 254.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-7700

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-7700
Availability details for
Quantity
  • Availability
  • Add to Cart