COM497511034 Combi-Blocks, Inc.

2-(Bromomethyl)-5-ethoxy-1,3-difluorobenzene

  • MDL number MFCD09258697
  • Cas Number : 1017779-51-9
  • Molecular Weight : 251.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7945

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7945
Availability details for
Quantity
  • Availability
  • Add to Cart