COM497511095 Combi-Blocks, Inc.

5-Methyl-4-nitro-1h-pyrazole-3-carboxylic acid

  • MDL number MFCD08456975
  • Cas Number : 5334-38-3
  • Molecular Weight : 171.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-0083

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-0083
Availability details for
Quantity
  • Availability
  • Add to Cart