COM497511120 Combi-Blocks, Inc.

Dl-2'-methylphenylalanine hydrochloride

  • MDL number MFCD01320672
  • Cas Number : 22888-51-3
  • Molecular Weight : 215.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-0254

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-0254
Availability details for
Quantity
  • Availability
  • Add to Cart