COM497511133 Combi-Blocks, Inc.

3-Methyl-4-nitro-1h-pyrazole-5-carboxylic acid

  • MDL number MFCD00458002
  • Cas Number : 5334-38-3
  • Molecular Weight : 171.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-0364

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-0364
Availability details for
Quantity
  • Availability
  • Add to Cart