COM497511135 Combi-Blocks, Inc.

(5-Methyl-1h-imidazol-4-yl)methanamine

  • MDL number MFCD09751153
  • Cas Number : 66780-83-4
  • Molecular Weight : 111.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-0397

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-0397
Availability details for
Quantity
  • Availability
  • Add to Cart