COM497511757 Combi-Blocks, Inc.

2-Chloro-4-methyloxazole

  • MDL number MFCD13189379
  • Cas Number : 1060816-10-5
  • Molecular Weight : 117.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-4416

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-4416
Availability details for
Quantity
  • Availability
  • Add to Cart