COM497511786 Combi-Blocks, Inc.

6-Hydrazinyl-7h-purine

  • MDL number MFCD00129734
  • Cas Number : 5404-86-4
  • Molecular Weight : 150.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-4593

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-4593
Availability details for
Quantity
  • Availability
  • Add to Cart