COM497511836 Combi-Blocks, Inc.

4-Pyridinylboronic acid mida ester

  • MDL number MFCD11215236
  • Cas Number : 1104636-72-7
  • Molecular Weight : 234.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-4858

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-4858
Availability details for
Quantity
  • Availability
  • Add to Cart