COM497512050 Combi-Blocks, Inc.

2-(Chloromethyl)-5-(trifluoromethyl)pyridine

  • MDL number MFCD10697590
  • Cas Number : 128790-14-7
  • Molecular Weight : 195.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-6084

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-6084
Availability details for
Quantity
  • Availability
  • Add to Cart