COM497512141 Combi-Blocks, Inc.

2-Mercapto-5-methylpyridine

  • MDL number MFCD02684159
  • Cas Number : 18368-58-6
  • Molecular Weight : 125.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-6575

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-6575
Availability details for
Quantity
  • Availability
  • Add to Cart