COM497512557 Combi-Blocks, Inc.

Methyl 5-formyl-2-furoate

  • MDL number MFCD06803263
  • Cas Number : 5904-71-2
  • Molecular Weight : 154.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-9372

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-9372
Availability details for
Quantity
  • Availability
  • Add to Cart