COM497512695 Combi-Blocks, Inc.

Quinoline-4,7-diol

  • MDL number MFCD09260461
  • Cas Number : 955938-89-3
  • Molecular Weight : 161.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-0400

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-0400
Availability details for
Quantity
  • Availability
  • Add to Cart