COM497513059 Combi-Blocks, Inc.

2,2-Difluoro-1,3-dimethylimidazolidine

  • MDL number MFCD03788478
  • Cas Number : 220405-40-3
  • Molecular Weight : 136.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-3188

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-3188
Availability details for
Quantity
  • Availability
  • Add to Cart