COM497513067 Combi-Blocks, Inc.

7-Methoxy-n-(3-(trifluoromethyl)phenyl)quinoline-4-carboxamide

  • MDL number MFCD26404046
  • Cas Number : 1624262-18-5
  • Molecular Weight : 346.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-3235

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-3235
Availability details for
Quantity
  • Availability
  • Add to Cart