COM497513163 Combi-Blocks, Inc.

4-Phenyl-1,2,3-thiadiazole-5-carboxylic acid

  • MDL number MFCD00052216
  • Cas Number : 78875-63-5
  • Molecular Weight : 206.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-3915

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-3915
Availability details for
Quantity
  • Availability
  • Add to Cart